Invitrogen
Manufacturer Code:PA570174
Catalog # PIPA570174
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N-terminal region of mouse Glycerol kinase |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ASTP; ATP-stimulated glucocorticoid-receptor translocation promoter; ATP-stimulated glucocorticoid-receptor translocaton promoter; ATP:glycerol 3-phosphotransferase; GK; glycerokinase; Glycerol kinase; Glycerol kinase EC 2.7.1.30 ATP:glycerol 3-phosphotransferase Glycerokinase GKl
Gene Aliases: ASTP; D930012N15Rik; GK; GK1; GKD; Gyk
UniProt ID: (Human) P32189, (Bovine) Q0IID9, (Rat) Q63060, (Mouse) Q64516
Entrez Gene ID: (Rabbit) 100354726, (Human) 2710, (Dog) 480872, (Bovine) 505987, (Horse) 100051016, (Guinea pig) 100726168, (Rat) 79223, (Mouse) 14933
Molecular Function:
carbohydrate kinase
kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.