Invitrogen
Manufacturer Code:MA518967
Catalog # PIMA518967
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3G4 |
Immunogen | GK2 (AAH29820 1 a.a. &bsim 553 a.a) full-length recombit protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ATP:glycerol 3-phosphotransferase 2; GK 2; GKP2 GKTA ATP:glycerol 3-phosphotransferase 2 GK 2 glycerokinase 2 glycerol kinase pseudogene 2 glycerol kinase testis specific 2 glycerol kinase testis specific 2; glycerokinase 2; Glycerol kinase 2; glycerol kinase pseudogene 2; glycerol kinase testis specific 2; Glycerol kinase, testis specific 2; testis tissue sperm-binding protein Li 77m
Gene Aliases: GK2; GKP2; GKTA
UniProt ID: (Human) Q14410
Entrez Gene ID: (Human) 2712
Molecular Function:
carbohydrate kinase
kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.