Invitrogen
Manufacturer Code:PA575254
Catalog # PIPA575254
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 276-325 of Human Glycerol kinase 2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP:glycerol 3-phosphotransferase 2; GK 2; glucokinase activity, related sequence 2; glycerokinase 2; Glycerol kinase 2; Glycerol kinase 2 GK 2 Glycerokinase 2 ATP:glycerol 3-phosphotransferase 2 Glycerol kinase testis specific 2 GK2 GKP2 GKTA; glycerol kinase pseudogene 2; glycerol kinase testis specific 2; Glycerol kinase, testis specific 2; testis tissue sperm-binding protein Li 77m
Gene Aliases: Gk-rs2; GK2; GKP2; Gkrs2; GKTA
UniProt ID: (Human) Q14410, (Mouse) Q9WU65
Entrez Gene ID: (Human) 2712, (Mouse) 14626, (Rat) 289481
Molecular Function:
carbohydrate kinase
kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.