Novus Biologicals
Manufacturer Code:NBP15703320UL
Catalog # NBP15703320
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to GK (glycerol kinase) The peptide sequence was selected from the middle region of GK. Peptide sequence MAAGAAEGVGVWSLEPEDLSAVTMERFEPQINAEESEIRYSTWKKAVMKS. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP:glycerol 3-phosphotransferase; ATP:glycerol 3-phosphotransferase EC 2.7.1.30 GK1 GKD Glycerokinase glycerol kinase; GK; glycerokinase; Glycerol kinase
Gene Aliases: GK; GK1; GKD
UniProt ID: (Human) P32189
Entrez Gene ID: (Human) 2710
Molecular Function:
carbohydrate kinase
kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.