Novus Biologicals
Manufacturer Code:NBP290308AF350
Catalog # NB072082
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 104 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Human Glycerol 3 Phosphate Dehydrogenase (P21695 Met1-Met349). |
Conjugate | Alexa Fluor 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: EC 1.1.1 EC 1.1.1.8 FLJ26652 glycerol-3-phosphate dehydrogenase [NAD+] cytoplasmic glycerol-3-phosphate dehydrogenase 1 (soluble) glycerol-3-phosphate dehydrogenase soluble GPD-C GPDH-C; glycerol-3-phosphate dehydrogenase 1 (soluble); Glycerol-3-phosphate dehydrogenase [NAD(+)], cytoplasmic; glycerol-3-phosphate dehydrogenase [NAD+], cytoplasmic; glycerophosphate dehydrogenase; GPD-C
Gene Aliases: GPD-C; GPD1; GPDH-C; HTGTI
UniProt ID: (Human) P21695
Entrez Gene ID: (Human) 2819
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.