Novus Biologicals
Manufacturer Code:NBP159657
Catalog # NBP159657
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to SLC6A5(solute carrier family 6 (neurotransmitter transporter glycine) member 5) The peptide sequence was selected from the middle region of SLC6A5. Peptide sequence LIVTCTNSATSIFAGFVIFSVIGFMANERKVNIENV |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GlyT-2; GlyT2 GLYT-2 GLYT2GlyT-2 NET1 sodium- and chloride-dependent glycine transporter 2 solute carrier family 6 (neurotransmitter transporter glycine) member 5 Solute carrier family 6 member 5; Sodium- and chloride-dependent glycine transporter 2; solute carrier family 6 (neurotransmitter transporter, glycine), member 5; Solute carrier family 6 member 5
Gene Aliases: GLYT-2; GLYT2; HKPX3; NET1; SLC6A5
UniProt ID: (Human) Q9Y345
Entrez Gene ID: (Human) 9152
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.