Invitrogen
Manufacturer Code:PA542437
Catalog # PIPA542437
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human SLC6A5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: glycine transporter 2; GlyT-2; slc6a5; Sodium- and chloride-dependent glycine transporter 2; Solute carrier family 6 (neurotransmitter transporter glycine) member 5 GLYT2 NET1 GLYT-2; solute carrier family 6 (neurotransmitter transporter), member 5; solute carrier family 6 (neurotransmitter transporter, glycine), member 5; Solute carrier family 6 member 5
Gene Aliases: GLYT-2; GLYT2; HKPX3; NET1; prestin; si:dkey-7n10.12; SLC6A5
UniProt ID: (Human) Q9Y345, (Rat) P58295
Entrez Gene ID: (Rabbit) 100339626, (Horse) 100057380, (Human) 9152, (Dog) 612095, (Rat) 171148, (Mouse) 104245, (Zebrafish) 494450
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.