Invitrogen
Manufacturer Code:PA577456
Catalog # PIPA577456
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)RLYVLKLSDDIGD corresponding to amino acid residues 202-214 of rat Glycine Transporter 1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: glycine transporter 1; glycine transporter variant 1a; GlyT-1; SLC6A9; Sodium- and chloride-dependent glycine transporter 1; solute carrier family 6 (neurotransmitter transporter, glycine), member 9; Solute carrier family 6 member 9
Gene Aliases: GLYT-1; GLYT-1b; GLYT1; SLC6A9
UniProt ID: (Human) P48067, (Mouse) P28571, (Rat) P28572
Entrez Gene ID: (Human) 6536, (Mouse) 14664, (Rat) 116509
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.