Invitrogen
Manufacturer Code:PA569166
Catalog # PIPA569166
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of mouse SLC6A9 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: glycine transporter 1; glycine transporter variant 1a; GlyT-1; Sodium- and chloride-dependent glycine transporter 1; solute carrier family 6 (neurotransmitter transporter, glycine), member 9; Solute carrier family 6 member 9
Gene Aliases: GLYT-1; GLYT-1b; GLYT1; SLC6A9
UniProt ID: (Bovine) Q28039, (Human) P48067, (Mouse) P28571, (Rat) P28572
Entrez Gene ID: (Horse) 100053016, (Dog) 482526, (Bovine) 282368, (Human) 6536, (Rabbit) 100351167, (Guinea pig) 100720563, (Mouse) 14664, (Rat) 116509
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.