Novus Biologicals
Manufacturer Code:NBP299590F
Catalog # NB061903
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human Glutathione Synthetase. |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 6.3.2.3 Glutathione synthase glutathione synthetase GSH synthetase GSHS GSH-S MGC14098; Glutathione synthase; Glutathione synthetase; GSH synthetase; GSH-S
Gene Aliases: GSHS; GSS; HEL-S-64p; HEL-S-88n
UniProt ID: (Human) P48637
Entrez Gene ID: (Human) 2937
Molecular Function: ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.