R&D Systems
Manufacturer Code:MAB6455SP
Catalog # MAB6455SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 800027 |
Immunogen | <EM>E. coli</EM>-derived recombinant human Glutathione S-Transferase pi 1/GSTP1 Met1-Glu210 Accession # AAC51280 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: deafness X-linked 7 DFN7 EC 2.5.1.18 FAEES3 fatty acid ethyl ester synthase III glutathione S-transferase P glutathione S-transferase pi 1 Glutathione STransferase pi 1 GST class-pi GST3DFN7 GSTP GSTP1 GSTP1-1 PI; deafness, X-linked 7; epididymis secretory protein Li 22; fatty acid ethyl ester synthase III; Glutathione S-transferase P; GST class-pi; GSTP1-1
Gene Aliases: DFN7; FAEES3; GST3; GSTP; GSTP1; HEL-S-22; PI
UniProt ID: (Human) P09211
Entrez Gene ID: (Human) 2950
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.