Novus Biologicals
Manufacturer Code:NBP297945AF488
Catalog # NB049708
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human Glutathione Reductase fragment. |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 1.8.1 EC 1.8.1.7 GLUR glutathione reductase glutathione reductase mitochondrial GR GRase GRD1 MGC78522; epididymis luminal protein 75; epididymis secretory sperm binding protein Li 122m; glutathione reductase; Glutathione reductase, mitochondrial; glutathione S-reductase; GR; GRase
Gene Aliases: GLUR; GRD1; GSR; HEL-75; HEL-S-122m
UniProt ID: (Human) P00390
Entrez Gene ID: (Human) 2936
Molecular Function: dehydrogenase oxidase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.