Invitrogen
Manufacturer Code:PA579316
Catalog # PIPA579316
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence in the middle region of human Glutaredoxin 2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bA101E13.1 (GRX2 glutaredoxin (thioltransferase) 2); glutaredoxin (thioltransferase) 2; glutaredoxin 2 (thioltransferase); Glutaredoxin-2 mitochondrial GLRX2 GRX2 CGI-133; Glutaredoxin-2, mitochondrial
Gene Aliases: 1700010P22Rik; AI645710; CGI-133; GLRX2; GRX2
UniProt ID: (Human) Q9NS18, (Mouse) Q923X4, (Rat) Q6AXW1
Entrez Gene ID: (Human) 51022, (Mouse) 69367, (Rat) 114022
Molecular Function:
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.