Novus Biologicals
Manufacturer Code:NBP300300F
Catalog # NB065469
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant Glutaredoxin 1/GLRX1 (P35754 Met1-Gln106). |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: glutaredoxin (thioltransferase); glutaredoxin (thioltransferase) glutaredoxin-1 GRXGRX1 MGC117407 Thioltransferase-1 TTase-1; Glutaredoxin-1; thioltransferase; Thioltransferase-1; TTase-1
Gene Aliases: GLRX; GRX; GRX1
UniProt ID: (Human) P35754
Entrez Gene ID: (Human) 2745
Molecular Function: oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.