R&D Systems
Manufacturer Code:AF3399
Catalog # AF3399
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human GLRX1 Met1-Gln106 Accession # P35754 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: GLRX glutaredoxin (thioltransferase) glutaredoxin-1 GRXGRX1 MGC117407 Thioltransferase Thioltransferase-1 TTase-1; glutaredoxin (thioltransferase); Glutaredoxin-1; thioltransferase; Thioltransferase-1; TTase-1
Gene Aliases: GLRX; GRX; GRX1
UniProt ID: (Human) P35754
Entrez Gene ID: (Human) 2745
Molecular Function:
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.