R&D Systems
Manufacturer Code:AF6368SP
Catalog # AF6368SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>S. frugiperda</EM> insect ovarian cell line <EM>Sf</EM> 21-derived recombinant human Glutaminyl-peptide Cyclotransferase/QPCT Ala33-Leu361 Accession # Q16769 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: EC; EC 2.3.2.5 GCT Glutaminyl cyclase glutaminyl-peptide cyclotransferase Glutaminyl-tRNA cyclotransferase Glutamyl cyclase QCEC sQC; Glutaminyl cyclase; Glutaminyl-peptide cyclotransferase; Glutaminyl-tRNA cyclotransferase; Glutamyl cyclase; QC
Gene Aliases: GCT; QC; QPCT; sQC
UniProt ID: (Human) Q16769
Entrez Gene ID: (Human) 25797
Molecular Function: transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.