Novus Biologicals
Manufacturer Code:NBP243826AF750
Catalog # NP243826A70
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 7711 |
Immunogen | Full length human Glutamine synthetase Recombinant protein. |
Conjugate | Alexa Fluor 750 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: cell proliferation-inducing protein 59; cell proliferation-inducing protein 59 EC 4.1.1.15 EC 6.3.1.2 GLNSPIG59 Glutamate decarboxylase glutamate-ammonia ligase Glutamate--ammonia ligase glutamate-ammonia ligase (glutamine synthase) glutamine synthase glutamine synthetase GS PIG43 proliferation-inducing protein 43; glutamate decarboxylase; Glutamate--ammonia ligase; glutamine synthase; Glutamine synthetase; GS; Palmitoyltransferase GLUL; proliferation-inducing protein 43
Gene Aliases: GLNS; GLUL; GS; PIG43; PIG59
UniProt ID: (Human) P15104
Entrez Gene ID: (Human) 2752
Molecular Function: ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.