Invitrogen
Manufacturer Code:PA581505
Catalog # PIPA581505
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to the C-terminus of the Human GLUL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cell proliferation-inducing protein 59; glutamate decarboxylase; Glutamate--ammonia ligase; Glutamate-ammonia ligase GS; glutamine synthase; Glutamine synthetase; GS; Palmitoyltransferase GLUL; proliferation-inducing protein 43
Gene Aliases: GLNS; GLUL; GS; PIG43; PIG59
UniProt ID: (Human) P15104
Entrez Gene ID: (Human) 2752
Molecular Function: ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.