Invitrogen
Manufacturer Code:PA586977
Catalog # PIPA586977
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein of human GLS. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GLS; glutaminase C; Glutaminase kidney isoform mitochondrial GLS K-glutaminase L-glutamine amidohydrolase GLS1 KIAA0838; Glutaminase kidney isoform, mitochondrial; Glutaminase kidney isoform, mitochondrial 65 kDa chain; Glutaminase kidney isoform, mitochondrial 68 kDa chain; glutaminase, phosphate-activated; K-glutaminase; L-glutamine amidohydrolase
Gene Aliases: 6330442B14; AAD20; AI314027; B230365M23Rik; GAC; GAM; GLS; GLS1; KGA; KIAA0838
UniProt ID: (Human) O94925, (Mouse) D3Z7P3
Entrez Gene ID: (Human) 2744, (Mouse) 14660
Molecular Function:
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.