Invitrogen
Manufacturer Code:PA585416
Catalog # PIPA585416
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the C-terminus region of human Glutaminase. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AAD20 antibody DKFZp686O15119 antibody FLJ10358 antibody GLS1 antibody KIAA0838 antibody GLS antibody glutaminase C antibody K-glutaminase antibody glutaminase phosphate-activated antibody glutaminase kidney isoform mitochondrial antibody L-glutamine amidohydrolase antibody glutaminase antibody; GLS; Glutaminase kidney isoform, mitochondrial; Glutaminase kidney isoform, mitochondrial 65 kDa chain; Glutaminase kidney isoform, mitochondrial 68 kDa chain
Gene Aliases: 6330442B14; AI314027; B230365M23Rik; Gls; Gls1; Kiaa0838
UniProt ID: (Mouse) D3Z7P3
Entrez Gene ID: (Mouse) 14660
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.