Novus Biologicals
Manufacturer Code:NBP271285PECY55
Catalog # N271285PEC5
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI9F3 |
Immunogen | Full length recombinant protein of human Glut5 (NP_003030) produced in HEK293T cells. |
Conjugate | PE-Cyanine5.5 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. Do not freeze. |
Mouse Monoclonal Antibody
Protein Aliases: Fructose transporter; Fructose transporter Glucose transporter type 5 small intestine GLUT-5 GLUT5glucose transporter-like protein 5 solute carrier family 2 (facilitated glucose transporter) member 5 solute carrier family 2 (facilitated glucose/fructose transporter) member 5 solute carrier family 2 facilitated glucose transporter member 5; Glucose transporter type 5, small intestine; glucose transporter-like protein 5; GLUT-5; solute carrier family 2 (facilitated glucose/fructose transporter), member 5; Solute carrier family 2, facilitated glucose transporter member 5; testicular tissue protein Li 81
Gene Aliases: GLUT-5; GLUT5; SLC2A5
UniProt ID: (Human) P22732
Entrez Gene ID: (Human) 6518
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.