R&D Systems
Manufacturer Code:MAB8654
Catalog # MAB8654
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 925951 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human Glut4 Met1-Asp509 Accession # P14672 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Glucose transporter type 4, insulin-responsive; GLUT-4; GLUT-4 GLUT4Glucose transporter type 4 insulin-responsive insulin-responsive glucose transporter type 4 SLC2A4 solute carrier family 2 (facilitated glucose transporter) member 4 solute carrier family 2 facilitated glucose transporter member 4; insulin-responsive glucose transporter type 4; solute carrier family 2 (facilitated glucose transporter), member 4; Solute carrier family 2, facilitated glucose transporter member 4
Gene Aliases: GLUT4; SLC2A4
UniProt ID: (Human) P14672
Entrez Gene ID: (Human) 6517
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.