Novus Biologicals
Manufacturer Code:NBP244298
Catalog # NBP244298
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 6H11NB |
Immunogen | Partial recombinant human Glut4 protein (between amino acids 300-350) [Uniprot: P14672] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Glucose transporter type 4, insulin-responsive; GLUT-4; GLUT-4 GLUT4Glucose transporter type 4 insulin-responsive insulin-responsive glucose transporter type 4 solute carrier family 2 (facilitated glucose transporter) member 4 solute carrier family 2 facilitated glucose transporter member 4; insulin-responsive glucose transporter type 4; solute carrier family 2 (facilitated glucose transporter), member 4; Solute carrier family 2, facilitated glucose transporter member 4
Gene Aliases: GLUT4; SLC2A4
UniProt ID: (Human) P14672
Entrez Gene ID: (Human) 6517
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.