Novus Biologicals
Manufacturer Code:NBP149840
Catalog # NBP149840
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-TELEYLGPDEND corresponding to C-Terminus according to NP_001033.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Glucose transporter type 4, insulin-responsive; GLUT-4; GLUT-4 GLUT4Glucose transporter type 4 insulin-responsive insulin-responsive glucose transporter type 4 solute carrier family 2 (facilitated glucose transporter) member 4 solute carrier family 2 facilitated glucose transporter member 4; insulin-responsive glucose transporter type 4; solute carrier family 2 (facilitated glucose transporter), member 4; Solute carrier family 2, facilitated glucose transporter member 4
Gene Aliases: GLUT4; SLC2A4
UniProt ID: (Human) P14672
Entrez Gene ID: (Human) 6517
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.