R&D Systems
Manufacturer Code:FAB1415R100UG
Catalog # FAB1415R100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 202017 |
Immunogen | NS0 mouse myeloma cell line transfected with human Glut3 Accession # P11169 |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 °C as supplied. |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: FLJ90380 GLUT-3 GLUT3Glucose transporter type 3 brain SLC2A3 solute carrier family 2 (facilitated glucose transporter) member 3 solute carrier family 2 facilitated glucose transporter member 3; Glucose transporter type 3, brain; GLUT-3; solute carrier family 2 (facilitated glucose transporter), member 3; Solute carrier family 2, facilitated glucose transporter member 3
Gene Aliases: GLUT3; SLC2A3
UniProt ID: (Human) P11169
Entrez Gene ID: (Human) 6515
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.