R&D Systems
Manufacturer Code:MAB1414SP
Catalog # MAB1414SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 199017 |
Immunogen | NS0 mouse myeloma cell line transfected with human Glut2 Met1-Val524 Accession # P11168 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Glucose transporter type 2, liver; GLUT-2; GLUT-2 GLUT2 Glucose transporter type 2 liver SLC2A2 solute carrier family 2 (facilitated glucose transporter) member 2 solute carrier family 2 facilitated glucose transporter member 2; solute carrier family 2 (facilitated glucose transporter), member 2; Solute carrier family 2, facilitated glucose transporter member 2
Gene Aliases: GLUT2; SLC2A2
UniProt ID: (Human) P11168
Entrez Gene ID: (Human) 6514
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.