Novus Biologicals
Manufacturer Code:NBP169466
Catalog # NBP169466
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to SLC2A2(solute carrier family 2 (facilitated glucose transporter) member 2) The peptide sequence was selected from the N terminal of human SLC2A2. Peptide sequence ITAVLGSFQFGYDIGVINAPQQVIISHYRHVLGVPLDDRKAINNY |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Glucose transporter type 2, liver; GLUT-2; GLUT-2 GLUT2 Glucose transporter type 2 liver SLC2A2 solute carrier family 2 (facilitated glucose transporter) member 2 solute carrier family 2 facilitated glucose transporter member 2; solute carrier family 2 (facilitated glucose transporter), member 2; Solute carrier family 2, facilitated glucose transporter member 2
Gene Aliases: GLUT2; SLC2A2
UniProt ID: (Human) P11168
Entrez Gene ID: (Human) 6514
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.