R&D Systems
Manufacturer Code:FAB1418G100UG
Catalog # FAB1418G100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 202915 |
Immunogen | NS0 mouse myeloma cell line transfected with human Glut1 Met1-Val492 Accession # AAA52571 |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: Choreoathetosis/Spasticity Episodic (Paroxysmal Choreoathetosis) CSE DYT17 DYT18 DYT9 EIG12 Glucose transporter type 1 erythrocyte/brain GLUT-1 GLUT1DS HepG2 glucose transporter HTLVR Human T-Cell Leukemia Virus (I and II) Receptor MGC141895 MGC141896 PED SLC2A1 solute carrier family 2 (facilitated glucose transporter) member 1 Solute Carrier Family 2 Member 1 solute carrier family 2 facilitated glucose transporter member 1; choreoathetosis/spasticity, episodic (paroxysmal choreoathetosis/spasticity); Glucose transporter type 1, erythrocyte/brain; GLUT-1; HepG2 glucose transporter; human T-cell leukemia virus (I and II) receptor; receptor for HTLV-1 and HTLV-2; solute carrier family 2 (facilitated glucose transporter), member 1; Solute carrier family 2, facilitated glucose transporter member 1
Gene Aliases: CSE; DYT17; DYT18; DYT9; EIG12; GLUT; GLUT-1; GLUT1; GLUT1DS; HTLVR; PED; SDCHCN; SLC2A1
UniProt ID: (Human) P11166
Entrez Gene ID: (Human) 6513
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.