Novus Biologicals
Manufacturer Code:NBP303337100UL
Catalog # NB092515
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human Glucuronosyltransferase 1A1/UGT1A1 (NP_000454.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bilirubin UDP-glucuronosyltranserase; bilirubin UDP-glucuronosyltransferase 1-1; bilirubin UDP-glucuronosyltransferase 1-1 bilirubin UDP-glucuronosyltransferase isozyme 1 Bilirubin-specific UDPGT isozyme 1 GNT1EC 2.4.1.17 HUG-BR1 UDP glucuronosyltransferase 1 family polypeptide A1 UDP glycosyltransferase 1 family polypeptide A1 UDP-glucuronosyltransferase 1-1 UDP-glucuronosyltransferase 1-A UDP-glucuronosyltransferase 1A1 UDPGT UGT1*1 UGT1.1 UGT1-01 UGT-1A UGT1AUDPGT 1-1 UGT1BILIQTL1; bilirubin UDP-glucuronosyltransferase isozyme 1; Bilirubin-specific UDPGT isozyme 1; hUG-BR1; UDP glucuronosyltransferase 1 family, polypeptide A1; UDP glycosyltransferase 1 family, polypeptide A1; UDP-glucuronosyltransferase 1-1; UDP-glucuronosyltransferase 1-A; UDP-glucuronosyltransferase 1A isoform 1; UDP-glucuronosyltransferase 1A1; UDPGT 1-1; UGT-1A; UGT1*1; UGT1-01; UGT1.1; UGT1A1
Gene Aliases: BILIQTL1; GNT1; HUG-BR1; UDPGT; UDPGT 1-1; UGT1; UGT1A; UGT1A1
UniProt ID: (Human) P22309
Entrez Gene ID: (Human) 54658
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.