R&D Systems
Manufacturer Code:AF6490
Catalog # AF6490
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human Glucuronosyltransferase 1A1/UGT1A1 Leu60-Thr168 Accession # P22309 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: bilirubin UDP-glucuronosyltranserase; bilirubin UDP-glucuronosyltransferase 1-1; bilirubin UDP-glucuronosyltransferase 1-1 bilirubin UDP-glucuronosyltransferase isozyme 1 Bilirubin-specific UDPGT isozyme 1 GNT1 GNT1EC 2.4.1.17 HUG-BR1 UDP glucuronosyltransferase 1 family polypeptide A1 UDP glycosyltransferase 1 family polypeptide A1 UDP-glucuronosyltransferase 1-1 UDP-glucuronosyltransferase 1-A UDP-glucuronosyltransferase 1A1 UDPGT UDPGT 1-1 UG-BR1 UGT1 UGT1*1 UGT1.1 UGT1-01 UGT1A UGT-1A UGT1AUDPGT 1-1 UGT1BILIQTL1; bilirubin UDP-glucuronosyltransferase isozyme 1; Bilirubin-specific UDPGT isozyme 1; hUG-BR1; UDP glucuronosyltransferase 1 family, polypeptide A1; UDP glycosyltransferase 1 family, polypeptide A1; UDP-glucuronosyltransferase 1-1; UDP-glucuronosyltransferase 1-A; UDP-glucuronosyltransferase 1A isoform 1; UDP-glucuronosyltransferase 1A1; UDPGT 1-1; UGT-1A; UGT1*1; UGT1-01; UGT1.1; UGT1A1
Gene Aliases: BILIQTL1; GNT1; HUG-BR1; UDPGT; UDPGT 1-1; UGT1; UGT1A; UGT1A1
UniProt ID: (Human) P22309
Entrez Gene ID: (Human) 54658
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.