Novus Biologicals
Manufacturer Code:NBP270803F
Catalog # NBP270803F
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4G4 |
Immunogen | Recombinant protein fragment corresponding to amino acids 40-315 of human Glucosylceramidase/GBA (NP_000148) produced in E.coli. |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Acid beta-glucosidase; Acid beta-glucosidase alglucerase Beta-glucocerebrosidase D-glucosyl-N-acylsphingosine glucohydrolase EC 3.2.1.45 GBA1 GC GCB GLUC glucosidase beta acid glucosidase beta acid (includes glucosylceramidase) glucosylceramidase imiglucerase lysosomal glucocerebrosidase; Alglucerase; Beta-GC; Beta-glucocerebrosidase; Cholesterol glucosyltransferase; Cholesteryl-beta-glucosidase; D-glucosyl-N-acylsphingosine glucohydrolase; glucosidase, beta, acid; glucosylceramidase-like protein; Imiglucerase; Lysosomal acid GCase; Lysosomal acid glucosylceramidase; lysosomal glucocerebrosidase; SGTase
Gene Aliases: GBA; GBA1; GC; GCB; GLUC
UniProt ID: (Human) P04062
Entrez Gene ID: (Human) 2629
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.