Novus Biologicals
Manufacturer Code:NB100236H
Catalog # NB100236H
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoblot (IB) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residues 50 and 100 of human Glucose-6-Phosphate Dehydrogenase using the numbering given in entry NP_000393.2 (GeneID 2539). |
Conjugate | HRP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G6PD; G6PD G6PD1EC 1.1.1.49 glucose-6-phosphate dehydrogenase; Glucose-6-phosphate 1-dehydrogenase
Gene Aliases: G6PD; G6PD1
UniProt ID: (Human) Q16000
Entrez Gene ID: (Human) 2539
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.