Novus Biologicals
Manufacturer Code:NBP131588
Catalog # NBP131588
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human H6PD. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 6-phosphogluconolactonase; 6-phosphogluconolactonase DKFZp686A01246 EC 1.1.1.49 EC 2.7.4.3 EC 3.1.1.31 G6PD H form G6PDH GDH/6PGL endoplasmic bifunctional protein GDHglucose 1- dehydrogenase glucose dehydrogenase glucose dehyrogenase glucose-6-phosphate dehydrogenase salivary hexose-6-phosphate dehydrogenase (glucose 1-dehydrogenase) MGC87643; 6PGL; G6PD, H form; GDH; GDH/6PGL endoplasmic bifunctional protein; glucose 1- dehydrogenase; Glucose 1-dehydrogenase; glucose dehydrogenase; glucose dehyrogenase; Glucose-6-phosphate dehydrogenase; glucose-6-phosphate dehydrogenase, salivary; Hexose-6-phosphate dehydrogenase; hexose-6-phosphate dehydrogenase (glucose 1-dehydrogenase)
Gene Aliases: CORTRD1; G6PDH; GDH; H6PD
UniProt ID: (Human) O95479
Entrez Gene ID: (Human) 9563
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.