Novus Biologicals
Manufacturer Code:NBP28978750UL
Catalog # NB021046
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 223 |
Immunogen | Recombinant Human Glucokinase/GCK Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: ATP:D-hexose 6-phosphotransferase; EC 2.7.1 EC 2.7.1.2 FGQTL3 GK GLK glucokinase (hexokinase 4) hexokinase-4 Hexokinase-D HK4pancreatic isozyme HKIV HXKP LGLK maturity onset diabetes of the young 2 MODY2; Glucokinase; glucokinase (hexokinase 4); hexokinase D, pancreatic isozyme; Hexokinase type IV; Hexokinase-4; Hexokinase-D; HK IV; HK4
Gene Aliases: FGQTL3; GCK; GK; GLK; HHF3; HK4; HKIV; HXKP; LGLK; MODY2
UniProt ID: (Human) P35557
Entrez Gene ID: (Human) 2645
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.