Invitrogen
Manufacturer Code:PA525382
Catalog # PIPA525382
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 236-262 amino acids from the central region of human NR3C1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GCCR GCR GR GRL glucocorticoid nuclear receptor variant 1 glucocorticoid receptor nuclear receptor subfamily 3 group C member 1 variant DL-1 nuclear receptor subfamily 3 group C member 1 variant NS-1; glucocorticoid nuclear receptor variant 1; Glucocorticoid receptor; glucocorticoid receptor 1; GR; Nuclear receptor subfamily 3 group C member 1; nuclear receptor subfamily 3, group C, member 1 (glucocorticoid receptor)
Gene Aliases: GCCR; GCR; GCRST; GR; GRL; Grl-1; Grl1; NR3C1
UniProt ID: (Human) Q53EP5
Entrez Gene ID: (Human) 2908, (Mouse) 14815
Molecular Function:
nuclear hormone receptor
nucleic acid binding
receptor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.