Invitrogen
Catalog # PA1-28366
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | 1:500-1:1,000 |
Species reactivity | Human, Non-human primate |
Host / Isotype | Sheep / IgG |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide consisting of amino acids 150-175 of squirrel monkey glucocorticoid receptor coupled to diptheria toxoid. This sequence is identical in human glucocorticoid receptor except for 2 amino acids (lowercase): N terminus - APkEKEFPKTHSDVSSEQQnLKGQTG-NH2 |
Conjugate | Unconjugated |
Form | Liquid |
Concentration | Conc. Not Determined |
Purification | purified |
Storage buffer | PBS |
Contains | no preservative |
Storage conditions | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
RRID | AB_1957682 |
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Glucocorticoid Receptor (NR3C1) is a receptor for glucocorticoids that can act as both a transcription factor and as a regulator of other transcription factors. Glucocorticoid Receptor can also be found in heteromeric cytoplasmic complexes along with heat shock factors and immunophilins. The protein is typically found in the cytoplasm until it binds a ligand, which induces transport into the nucleus. Glucocorticoid Receptor is expressed in the heart, detected in left and right atria, left and right ventricles, aorta, apex, intraventricular septum, and atrioventricular node as well as whole adult and fetal heart. Alternate splicing, the use of at least three different promoters, and alternate translation initiation sites result in several transcript variants encoding the same protein or different isoforms, but the full-length nature of some variants has not been determined. Mutations in the Glucocorticoid Receptor gene are a cause of glucocorticoid resistance, or cortisol, resistance. Studies have shown that glucocorticoid receptor (GR) must be associated with a complex of chaperone proteins for ligand activation. GR binds to known steroids such as dexamethasone with nanomolar affinity
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: glucocorticoid nuclear receptor variant 1; Glucocorticoid receptor; GR; gr alpha; GR Kit; Nuclear receptor subfamily 3 group C member 1; nuclear receptor subfamily 3, group C, member 1 (glucocorticoid receptor); OTTHUMP00000222854; OTTHUMP00000222858
Gene Aliases: GCCR; GCR; GCRST; GR; GRL; NR3C1
UniProt ID: (Human) P04150
Entrez Gene ID: (Human) 2908
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.