Invitrogen
Manufacturer Code:PA110856
Catalog # 01-671-897
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues 150-175 of squirrel monkey glucocorticoid receptor conjugated to diphtheria toxoid. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: glucocorticoid nuclear receptor variant 1; Glucocorticoid receptor; GR; GR GCCR GCR Glucocorticoid receptor lymphocyte GRL Grl1 NR3C1 Nuclear receptor subfamily 3 group C member 1; Nuclear receptor subfamily 3 group C member 1; nuclear receptor subfamily 3, group C, member 1 (glucocorticoid receptor)
Gene Aliases: GCCR; GCR; GCRST; GR; GRL; NR3C1
UniProt ID: (Human) Q53EP5
Entrez Gene ID: (Human) 2908
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.