Invitrogen
Manufacturer Code:PA577446
Catalog # PIPA577446
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)KTRYSQKIGDDLS corresponding to amino acid residues 200-212 of rat Glucagon Receptor |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GL-R; Glucagon Receptor; glucagon receptor perhaps same as Niddm3
Gene Aliases: GCGR; GGR; GL-R; GR
UniProt ID: (Human) P47871, (Mouse) Q61606, (Rat) P30082
Entrez Gene ID: (Human) 2642, (Mouse) 14527, (Rat) 24953
Molecular Function: G-protein coupled receptor antibacterial response protein defense/immunity protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.