Novus Biologicals
Manufacturer Code:NBP281065DL594
Catalog # NB015236
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | hGR-2 F6 |
Immunogen | Recombinant human Glucagon R/GCGR. |
Conjugate | DyLight 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: FLJ97182 GGR GL-R glucagon receptor MGC138246; GL-R; Glucagon receptor
Gene Aliases: GCGR; GGR; GL-R
UniProt ID: (Human) P47871
Entrez Gene ID: (Human) 2642
Molecular Function: G-protein coupled receptor antibacterial response protein defense/immunity protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.