Invitrogen
Manufacturer Code:14974380
Catalog # 14-974-380
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | ICACLS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: GCG Glicentin Glicentin-related polypeptide GLP1 GLP-1 GLP-1(7-36) GLP-1(7-37) GLP1/2 GLP2 GLP-2 Glu Glucagon glucagon-like peptide 1 Glucagon-like peptide 1(7-36) Glucagon-like peptide 1(7-37) Glucagon-like peptide 2 glucagon-like peptide-1 GRPP IL12p35 IL12P40 Incretin hormone OXM OXY Oxyntomodulin porcine proglucagon PPG preproglucagon proglucagon; Glicentin; Glicentin-related polypeptide; GLP-1; GLP-1(7-36); GLP-1(7-37); GLP-2; Glucagon; Glucagon-like peptide 1; Glucagon-like peptide 1(7-36); Glucagon-like peptide 1(7-37); Glucagon-like peptide 2; glucagon-like peptide-1; GRPP; Incretin hormone; OXM; Oxyntomodulin; preproglucagon; Pro-glucagon
Gene Aliases: GCG; GLP-1; GLP1; GLP2; Glu; GRPP; PPG
UniProt ID: (Human) P01275, (Mouse) P55095
Entrez Gene ID: (Human) 2641, (Mouse) 14526, (Rat) 24952
Molecular Function:
peptide hormone
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.