Invitrogen
Manufacturer Code:PA520450
Catalog # PIPA520450
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 13 amino acid peptide near the amino terminus of the human Grik3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dJ1090M5.1 (glutamate receptor, ionotropic, kainate 3 (GLUR7)); EAA5; EAA5 GLR7 GLUR7 GluR7a dJ1090M5.1 (glutamate receptor ionotropic kainate 3 (GLUR7)) excitatory amino acid receptor 5 gluR-7 glutamate receptor 7 glutamate receptor ionotropic kainate 3; Excitatory amino acid receptor 5; GluK3; GluR-7; Glutamate receptor 7; Glutamate receptor ionotropic, kainate 3; glutamate receptor subunit kainate subtype; glutamate receptor, ionotropic, kainate 3
Gene Aliases: 9630027E11; EAA5; GLR7; GluK3; Glur-7; GLUR7; GluR7-3; GluR7a; GRIK3
UniProt ID: (Human) Q13003, (Rat) P42264, (Mouse) B1AS29
Entrez Gene ID: (Human) 2899, (Rat) 298521, (Mouse) 14807
Molecular Function:
ion channel
ionotropic glutamate receptor
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.