Invitrogen
Manufacturer Code:PA518964
Catalog # PIPA518964
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (KIRQLPIDSDDSRP) corresponding to the internal amino acids of GRIK3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: GluK3; GluR-7; Glutamate receptor 7; Glutamate receptor ionotropic, kainate 3; glutamate receptor subunit kainate subtype; GRIK3 GLUR7 glutamate receptor ionotropic kainate 3 EAA5 GLR7 GluR7a dJ1090M5.1 (glutamate receptor ionotropic kainate 3 (GLUR7)) excitatory amino acid receptor 5 glutamate receptor 7
Gene Aliases: 9630027E11; GluK3; Glur-7; GluR7; GluR7-3; Grik3
UniProt ID: (Rat) P42264, (Mouse) B1AS29
Entrez Gene ID: (Rat) 298521, (Mouse) 14807
Molecular Function:
ion channel
ionotropic glutamate receptor
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.