Invitrogen
Manufacturer Code:PA137780
Catalog # 01-673-519
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide from C-terminus of human GluR6/7. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bA487F5.1; dJ1090M5.1 (glutamate receptor, ionotropic, kainate 3 (GLUR7)); EAA4; EAA5; Excitatory amino acid receptor 4; Excitatory amino acid receptor 5; GluK2; GluK3; GluR-6; GluR-7; Glutamate receptor 6; Glutamate receptor 7; glutamate receptor form A; glutamate receptor form B; glutamate receptor form C; glutamate receptor form D; glutamate receptor form E; Glutamate receptor ionotropic, kainate 2; Glutamate receptor ionotropic, kainate 3; glutamate receptor, ionotropic, kainate 2; glutamate receptor, ionotropic, kainate 3; GRIK2 GRIK3 EAA4 GLR6 MRT6 GLUK6 GLUR6 GluK2 Glutamate Receptor 6/7 Glutamate Receptor 6 Glutamate Receptor 7
Gene Aliases: EAA4; EAA5; GLR6; GLR7; GluK2; GluK3; GLUK6; GLUR6; GLUR7; GluR7a; GRIK2; GRIK3; MRT6
UniProt ID: (Human) Q13002, (Human) Q13003, (Rat) P42260
Entrez Gene ID: (Human) 2898, (Human) 2899, (Rat) 54257
Molecular Function:
ion channel
ionotropic glutamate receptor
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.