Invitrogen
Manufacturer Code:PA577418
Catalog # PIPA577418
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide CSAMVEELRMSLK corresponding to amino acid residues 858-870 of rat Kainate Receptor GluR6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bA487F5.1; EAA4; Excitatory amino acid receptor 4; GluK2; gluR beta-2; GluR-6; Glutamate receptor 6; glutamate receptor beta-2; glutamate receptor form A; glutamate receptor form B; glutamate receptor form C; glutamate receptor form D; glutamate receptor form E; Glutamate receptor ionotropic, kainate 2; glutamate receptor, ionotropic kainate 2; glutamate receptor, ionotropic, kainate 2; GRIK2
Gene Aliases: AW124492; EAA4; GLR6; GluK2; GLUK6; Glur-6; GLUR6; Glurbeta2; GRIK2; MRT6
UniProt ID: (Human) Q13002, (Rat) P42260
Entrez Gene ID: (Human) 2898, (Rat) 54257, (Mouse) 14806
Molecular Function:
ion channel
ionotropic glutamate receptor
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.