Novus Biologicals
Manufacturer Code:NBP176849
Catalog # NBP176849
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of the human Grik1. The immunogen is located within the last 50 amino acids of Grik1. Amino Acid Squence: CGKLIREERGIRKQS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EAA3; EEA3 GLR5 GluR5 GluR-5 glutamate receptor ionotropic kainate 1 ionotropic kainate 1; Excitatory amino acid receptor 3; GluK1; GluR-5; Glutamate receptor 5; Glutamate receptor ionotropic, kainate 1; glutamate receptor, ionotropic, kainate 1
Gene Aliases: EAA3; EEA3; GLR5; GluK1; gluR-5; GLUR5; GRIK1
UniProt ID: (Human) P39086
Entrez Gene ID: (Human) 2897
Molecular Function:
ion channel
ionotropic glutamate receptor
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.