Invitrogen
Manufacturer Code:PA518931
Catalog # PIPA518931
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (KKLDQREYPGSETP) corresponding to the internal amino acids of GRIA4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: AMPA-selective glutamate receptor 4; GluA4; GluR-4; GluR-D; Glutamate receptor 4; Glutamate receptor ionotropic, AMPA 4; glutamate receptor, ionotrophic, AMPA 4; glutamate receptor, ionotropic, AMPA 4; GRIA4 glutamate receptor ionotrophic AMPA 4 GLUR4 GLUR4C GLURD AMPA-selective glutamate receptor 4
Gene Aliases: GluA4; GLUR4; GLUR4C; GLURD; GRIA4
UniProt ID: (Human) P48058
Entrez Gene ID: (Human) 2893
Molecular Function:
ion channel
ionotropic glutamate receptor
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.