Invitrogen
Manufacturer Code:PA5116603
Catalog # PIPA5116603
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 254-433 of human GRIA4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AMPA-selective glutamate receptor 4; AMPA-selective glutamate receptor 4 GluA4 GluR4 Glur-4 GLUR4C Gluralpha4 GLURD gluR-D glutamate ionotropic receptor AMPA type subunit 4 glutamate receptor glutamate receptor 4 glutamate receptor channel alpha 4 glutamate receptor ionotropic AMPA 4 glutamate receptor ionotrophic AMPA 4 glutamate receptor ionotropic AMPA 4 glutamate receptor ionotropic AMPA4 (alpha 4) GRIA4 spike wave 1 spkw1; GluA4; GluR-4; GluR-D; Glutamate receptor 4; glutamate receptor channel alpha 4; Glutamate receptor ionotropic, AMPA 4; glutamate receptor, ionotrophic, AMPA 4; glutamate receptor, ionotropic, AMPA 4; glutamate receptor, ionotropic, AMPA4 (alpha 4); spike wave 1
Gene Aliases: GluA4; Glur-4; GluR-D; GLUR4; GLUR4C; Gluralpha4; GLURD; GRIA4; spkw1
UniProt ID: (Human) P48058, (Mouse) Q9Z2W8, (Rat) P19493
Entrez Gene ID: (Human) 2893, (Mouse) 14802, (Rat) 29629
Molecular Function: ion channel ionotropic glutamate receptor ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.