Invitrogen
Manufacturer Code:MA515869
Catalog # PIMA515869
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1D2 |
Immunogen | Purified recombit fragment of human GRIA3 expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AMPA-selective glutamate receptor 3; dJ1171F9.1; GluA3; GluR-3; GluR-C; GluR-K3; Glutamate receptor 3; glutamate receptor C; Glutamate receptor ionotropic, AMPA 3; glutamate receptor subunit 3; glutamate receptor, ionotrophic, AMPA 3; glutamate receptor, ionotropic, AMPA 3; RP11-349N19.3 GLUR-C GLUR-K3 GLUR3 GLURC GluA3 MRX94 AMPA-selective glutamate receptor 3 dJ1171F9.1 gluR-3 glutamate receptor 3 glutamate receptor C glutamate receptor ionotropic AMPA 3 glutamate receptor subunit 3
Gene Aliases: GluA3; GLUR-C; GLUR-K3; GLUR3; GLURC; GRIA3; MRX94
UniProt ID: (Human) P42263
Entrez Gene ID: (Human) 2892
Molecular Function:
ion channel
ionotropic glutamate receptor
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.