Invitrogen
Manufacturer Code:PA538105
Catalog # PIPA538105
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from human GluR2/3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AMPA selective glutamate receptor; AMPA-selective glutamate receptor 2; AMPA-selective glutamate receptor 2 GluR-2 GluR-B GluR-K2 GLUR2 GLURB Glutamate receptor 2 Glutamate receptor ionotropic AMPA 2 glutamate receptor ionotropic AMPA 2 GRIA2 HBGR2 GPRC1C GRM3 MGR3 Metabotropic glutamate receptor 3 precursor glutamate receptor metabotropic type 3; AMPA-selective glutamate receptor 3; dJ1171F9.1; GluA2; GluA3; GluR-2; GluR-3; GluR-B; GluR-C; GluR-K2; GluR-K3; Glutamate receptor 2; Glutamate receptor 3; glutamate receptor B; glutamate receptor B flip isoform; glutamate receptor C; glutamate receptor channel alpha3 subunit; Glutamate receptor ionotropic, AMPA 2; Glutamate receptor ionotropic, AMPA 3; glutamate receptor subunit 3; glutamate receptor, ionotrophic, AMPA 3; glutamate receptor, ionotropic, AMPA 2; glutamate receptor, ionotropic, AMPA 3; glutamate receptor, ionotropic, AMPA3 (alpha 3)
Gene Aliases: 2900064I19Rik; GluA2; GluA3; Glur-2; GluR-3; GluR-B; GLUR-C; GluR-K2; GLUR-K3; GLUR2; GLUR3; Gluralpha3; GLURB; GLURC; GRIA2; GRIA3; HBGR2; Kiaa4184; MRX94
UniProt ID: (Human) P42262, (Human) P42263, (Rat) P19491, (Mouse) Q9Z2W9, (Rat) P19492
Entrez Gene ID: (Human) 2891, (Human) 2892, (Mouse) 14800, (Rat) 29627, (Mouse) 53623, (Rat) 29628
Molecular Function: ion channel ionotropic glutamate receptor ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.