Invitrogen
Manufacturer Code:PA137773
Catalog # 01-673-512
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide from C-terminus of human GluR2/3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C do not freeze,do not freeze |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AMPA-selective glutamate receptor 2; AMPA-selective glutamate receptor 3; dJ1171F9.1; GluA2; GluA3; GluR-2; GluR-3; GluR-B; GluR-C; GluR-K2; GluR-K3; Glutamate receptor 2; Glutamate receptor 2 GluR-2 AMPA-selective glutamate receptor 2 GluR-B GluR-K2 Glutamate receptor ionotropic AMPA 2 GluA2 GRIA2 GLUR2; Glutamate receptor 3; glutamate receptor B flip isoform; glutamate receptor C; Glutamate receptor ionotropic, AMPA 2; Glutamate receptor ionotropic, AMPA 3; glutamate receptor subunit 3; glutamate receptor, ionotrophic, AMPA 3; glutamate receptor, ionotropic, AMPA 2; glutamate receptor, ionotropic, AMPA 3
Gene Aliases: GluA2; GluA3; GLUR-C; GluR-K2; GLUR-K3; GLUR2; GLUR3; GLURB; GLURC; GRIA2; GRIA3; HBGR2; MRX94
UniProt ID: (Human) P42262, (Human) P42263
Entrez Gene ID: (Human) 2891, (Human) 2892
Molecular Function:
ion channel
ionotropic glutamate receptor
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.